Skip to Content
Merck
All Photos(1)

Key Documents

SRP4388

Sigma-Aldrich

SDF-1alpha (CXCL12) from mouse

recombinant, expressed in E. coli, ≥99% (SDS-PAGE), ≥99% (HPLC)

Synonym(s):

CXCL12, PBSF, Pre-B cell growth-stimulating factor, SDF-1, chemokine (C-X-C motif) ligand 12, hIRH

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥99% (HPLC)
≥99% (SDS-PAGE)

form

lyophilized

potency

50-100 ng/mL

mol wt

~7.9 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Sdf1(20315)

General description

Stromal Cell-Derived Factor-1α (SDF-1α) is a recently discovered protein belonging to the α-chemokine (C-X-C) family of cytokines. Recombinant mouse SDF-1α is a 7.9kDa protein containing 68 amino acids residues.

Biochem/physiol Actions

Stromal Cell-Derived Factor-1α (SDF-1α) is a pleiotropic chemokine. It works as a chemoattractant for T- and B-lymphocytes and inflammatory monocytes. In neural stem cells, SDF-1α is needed for proliferation, survival, migration, intercellular interaction and guidance to neuronal axon. It is responsible for pathogenic inflammation in the central nervous system (CNS). It also controls BMP (bone morphogenic protein)-associated osteogenic differentiation.

Physical form

Lyophilized from 10 mM acetic acid. The protein may appear as a haze or film, which is difficult to see at the bottom of the vial.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitution in water to a concentration of 0.1-1.0 mg/mL. The solution can then be diluted into other aqueous buffers.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of the alpha-chemokine stromal cell-derived factor (SDF-1) in the developing and mature central nervous system.
Lazarini F, et al.
Glia, 42, 139-148 (2003)
Stephen Ryan Mut et al.
Micromachines, 13(3) (2022-03-27)
Millions of adults are affected by progressive vision loss worldwide. The rising incidence of retinal diseases can be attributed to damage or degeneration of neurons that convert light into electrical signals for vision. Contemporary cell replacement therapies have transplanted stem
BMP2 Regulation of CXCL12 Cellular, Temporal, and Spatial Expression is Essential During Fracture Repair.
Myers TJ, et al.
Journal of Bone and Mineral Research, 30, 2014-2027 (2015)
Rina Zilkha-Falb et al.
Journal of neuroinflammation, 13, 7-7 (2016-01-10)
Demyelination and axonal degeneration, hallmarks of multiple sclerosis (MS), are associated with the central nervous system (CNS) inflammation facilitated by C-X-C motif chemokine 12 (CXCL12) chemokine. Both in MS and in experimental autoimmune encephalomyelitis (EAE), the deleterious CNS inflammation has
Szandra Schwarcz et al.
Molecular biology reports, 50(6), 5273-5282 (2023-05-05)
Commensal bacteria secrete metabolites that reach distant cancer cells through the circulation and influence cancer behavior. Deoxycholic acid (DCA), a hormone-like metabolite, is a secondary bile acid specifically synthesized by intestinal microbes. DCA may have both pro- and antineoplastic effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service