Skip to Content
Merck
All Photos(1)

Key Documents

SML3413

Sigma-Aldrich

ASC-J9

≥98% (HPLC)

Synonym(s):

(1E, 4Z, 6E)-5-Hydroxy-1,7-bis(3,4-dimethoxyphenyl)-1,4,6-heptatrien-3-one, Dimethylcurcumin, GO-Y025

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24O6
CAS Number:
Molecular Weight:
396.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

, White to light orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

OC(/C=C/C1=CC(OC)=C(C=C1)OC)=C\C(/C=C/C2=CC(OC)=C(C=C2)OC)=O

InChI

1S/C23H24O6/c1-26-20-11-7-16(13-22(20)28-3)5-9-18(24)15-19(25)10-6-17-8-12-21(27-2)23(14-17)29-4/h5-15,24H,1-4H3/b9-5+,10-6+,18-15-

InChI key

ZMGUKFHHNQMKJI-CIOHCNBKSA-N

Biochem/physiol Actions

ASC-J9 (dimethylcurcumin) is an androgen receptor (AR)-selective degradation promoter that disrupts the interaction between AR and its coregulators. ASC-J9 induces AR degration in cultures (5 µM, LNCaP) and shows in vivo therapeutic efficacy (50 mg/kg i.p. qod) in murine bladder cancer (TCC-SUP and UMUC3 xenografts) and a spinal and bulbar muscular atrophy disease model (AR-polyQ transgenic SBMA mice).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Tian et al.
Journal of experimental & clinical cancer research : CR, 40(1), 3-3 (2021-01-05)
Early studies indicated that ASC-J9®, an androgen receptor (AR) degradation enhancer, could suppress the prostate cancer (PCa) progression. Here we found ASC-J9® could also suppress the PCa progression via an AR-independent mechanism, which might involve modulating the tumor suppressor ATF3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service