SML2026
TUG-1387
≥98% (HPLC)
Synonym(s):
4-Methyl-N-9H-xanthen-9-yl-benzamide, NSC 31078, NSC 31225
About This Item
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear (warmed)
shipped in
ambient
storage temp.
2-8°C
SMILES string
O=C(C1=CC=C(C)C=C1)NC2C3=C(C=CC=C3)OC4=C2C=CC=C4
InChI
1S/C21H17NO2/c1-14-10-12-15(13-11-14)21(23)22-20-16-6-2-4-8-18(16)24-19-9-5-3-7-17(19)20/h2-13,20H,1H3,(H,22,23)
InChI key
RWLHMQMVSKQATK-UHFFFAOYSA-N
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service