Skip to Content
Merck
All Photos(1)

Key Documents

SML0736

Sigma-Aldrich

Dafadine A

≥98% (HPLC)

Synonym(s):

4-[1-({ 5-[(2,6-Dimethylphenoxy)methyl]-3-isoxazolyl}carbonyl)-4-piperidinyl]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25N3O3
CAS Number:
Molecular Weight:
391.46
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

N3(CCC(CC3)c4ccncc4)C(=O)c1n[o]c(c1)COc2c(cccc2C)C

InChI

1S/C23H25N3O3/c1-16-4-3-5-17(2)22(16)28-15-20-14-21(25-29-20)23(27)26-12-8-19(9-13-26)18-6-10-24-11-7-18/h3-7,10-11,14,19H,8-9,12-13,15H2,1-2H3

InChI key

WNSNPYIHDMIFOO-UHFFFAOYSA-N

Biochem/physiol Actions

Dafadine A is a selective inhibitor of C. elegans DAF-9 cytochrome P450, a key regulator of dauer formation, developmental timing and longevity. C. elegans has four life stages. In harsh environmental conditions, nematodes can convert to the L3 stage, known as the ""non-aging"" dauer stage, in which development ceases until improved conditions are sensed. Dafadine A can induce dauer formation and increased longevity in wild-type worms raised in nonstressful conditions. Dafadine A is the only inhibitor known to robustly inhibit DAF-9 in wild-type worms. It does not inhibit DAF-12 activity. Dafadine is active in mammalian systems, inhibiting CYP27A1, the mammalian functional ortholog of DAF-9 that oxidizes 4-cholesten-3-one to make 26-hydroxy-4-cholesten-3-one and Δ4-dafachronic acid, without inhibiting other sterol- and oxysterol-metabolizing P450s such as CYP7A1 or CYP7B1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangxu Ma et al.
PLoS pathogens, 15(7), e1007960-e1007960 (2019-07-25)
Here, we discovered an endogenous dafachronic acid (DA) in the socioeconomically important parasitic nematode Haemonchus contortus. We demonstrate that DA promotes larval exsheathment and development in this nematode via a relatively conserved nuclear hormone receptor (DAF-12). This stimulatory effect is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service