SAB5702233
Anti-Cytokeratin 18 (KRT18) Antibody, clone 7B2N3, Rabbit Monoclonal
Synonym(s):
CK18, K18, Krt1-18
About This Item
Recommended Products
biological source
rabbit
Quality Level
material
colorless
clone
7B2N3, monoclonal
form
liquid
mol wt
48 kDa
species reactivity
rat, rat, human
concentration
0.8 mg/mL
technique(s)
immunofluorescence: 1:50 - 1:200
immunohistochemistry: 1:50 - 1:200
western blot: 1:1000 - 1:3000
color
colorless
General description
Immunogen
Application
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service