Skip to Content
Merck
All Photos(1)

Documents

SAB4301022

Sigma-Aldrich

Anti-MADCAM1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

MACAM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

human, mouse

concentration

0.6 mg/mL

technique(s)

western blot: 1:200-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_570116.2

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MADCAM1(8174)

Related Categories

Specificity

The antibody detects endogenous levels of total MADCAM1 protein.

Immunogen

Synthetic peptide corresponding to a region derived from internal residues of human mucosal vascular addressin cell adhesion molecule 1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service