Skip to Content
Merck
All Photos(1)

Key Documents

MAK247

Sigma-Aldrich

Sodium Assay Kit (Colorimetric)

sufficient for 100 colorimetric tests

Synonym(s):

Colorimetric Sodium Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

application(s)

cosmetics
food and beverages

detection method

colorimetric

storage temp.

−20°C

General description

Sodium (Na) is one of the most important electrolytes along with chloride, calcium and potassium. Na plays vital roles in the maintenance of plasma volume, pH balance, transmission of nerve impulses, and normal cell functions. Healthy individuals can absorb sodium ingested in food, and kidneys maintain proper sodium balance by excreting its excess in urine. Sodium sources include table salt, milk, meat, shellfish, bread, snack food, etc. Increased dietary sodium intake increases the risk of susceptibility to cardiovascular disease (CVD), most importantly with elevated blood pressure (BP).

Suitability

Suitable for the determination of sodium concentration in a variety of samples.

Principle

This sodium assay kit offers a simple, two-step colorimetric assay that is based on the requirement of sodium ion as a cofactor for the enzymatic activity of b-galactosidase (βG). Endogenous mono, di, and trivalent ions, ascorbic acid, creatinine, glucose, urea and bilirubin do not interfere with the assay. The kit can detect sodium ion concentrations as low as 25mM in a variety of samples.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sodium intake, blood pressure, and dietary sources of sodium in an adult South Indian population.
Ravi S, et al.
Annals of global health, 82(2), 234-242 (2016)
Study the Relationship between the Concentration of Minerals and Joints Pain.
Al-ghaliBi M J M.
Journal of Chemical and Pharmaceutical Research, 9(3) (2016)
The major electrolytes: sodium, potassium, and chloride.
Terry J.
Journal of Intravenous Nursing : The Official Publication of the Intravenous Nurses Society, 17(5), 240-247 (1994)
James T Grist et al.
Kidney international, 98(5), 1174-1178 (2020-06-26)
Sodium magnetic resonance imaging (MRI) is a powerful, non-invasive technique to assess sodium distribution within the kidney. Here we undertook pre-clinical and clinical studies to quantify the corticomedullary sodium gradient in healthy individuals and in a porcine model of diuresis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service