MAK244
Plasmin Activity Assay Kit (Fluorometric)
sufficient for 100 fluorometric tests
Synonym(s):
Plasmin Fluorometric Assay Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
detection method
fluorometric
relevant disease(s)
cancer; hematological disorder
storage temp.
−20°C
General description
Plasmin (EC 3.4.21.7) is a serine protease occurring in plasma as plasminogen. Upon activation via cleavage by plasminogen activators; plasmin solubilizes fibrin clots and activates and/or degrades compounds of the coagulation and complement systems. Plasminogen-Plasmin system has also been implicated in a wide variety of physiologic and pathologic processes, including tumor growth, invasion and metastasis.
Suitability
Suitable to determine the activity of pure plasmin and to detect the activity of plasmin in plasma.
Principle
The Plasmin Activity Assay Kit is based on the ability of plasmin to proteolytically cleave a synthetic substrate and release a fluorophore, AMC, which can be easily quantified by fluorescence microplate readers. This assay kit is simple, rapid, and can detect plasmin activity as low as 10 ng in a variety of samples.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Goss N.
Production of Plasma Proteins for Therapeutic Use, 2012, 496 -496 null
Kwaan H C and McMahon B
Coagulation in Cancer, 43-66 (2009)
Does plasmin have anticoagulant activity?.
Hoover-Plow J.
Vascular Health, 6, 199-199 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service