Skip to Content
Merck
All Photos(2)

Key Documents

MAK189

Sigma-Aldrich

A-KG Dehydrogenase Activity Assay Kit

Sufficient for 100 Colorimetric tests

Synonym(s):

α-KG Dehydrogenase Activity Assay Kit, α-Ketoglutarate Dehydrogenase Activity Colorimetric Assay Kit, AKGDH Assay Kit, Alpha-Ketoglutarate Dehydrogenase Activity Colorimetric Assay Kit, OGDC Activity Assay Kit, Oxoglutarate Dehydrogenase Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; orthopedic diseases; neurological disorders; rheumatological diseases; cancer; cardiovascular diseases

storage temp.

−20°C

Gene Information

human ... OGDH(4967)
mouse ... OGDH(18293)
rat ... OGDH(360975)

General description

α-Ketoglutarate Dehydrogenase (α-KGDH, EC 1.2.4.2) is a highly regulated Krebs cycle enzyme, forms the E1 subunit of the mitochondrial 2-oxoglutarate dehydrogenase complex. It catalyzes the conversion of α-ketoglutarate to succinyl-CoA and produces NADH directly providing electrons for the respiratory chain. Decreased α-KGDH complex activity has been associated with oxidative stress and neurodegenerative disorders such as Alzheimer′s and Parkinson′s diseases.

Application

α-Ketoglutarate Dehydrogenase Activity Colorimetric Assay Kit has been used to measure the enzyme activity of α-ketoglutarate dehydrogenase in samples.

Suitability

Suitable for the detection of α-ketoglutarate dehydrogenase activity in samples such as tissue, cells, and purified mitochondria.

Principle

The α-Ketoglutarate Dehydrogenase Activity Colorimetric Assay Kit provides a simple procedure for measuring α-ketoglutarate dehydrogenase activity in a variety of tissues, cells, and isolated mitochondria. α-KGDH activity is determined by measuring a colorimetric product with absorbance at 450 nm (A450) proportional to the enzymatic activity present. One unit of α-ketoglutarate dehydrogenase is the amount of enzyme that generates 1.0 μmole of NADH per minute at pH 7.5 at 37 °C.

Kit Components Only

Product No.
Description

  • KGDH Assay Buffer

  • KGDH Substrate

  • KGDH Developer

  • NADH Standard

  • KGDH Positive Control

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poldip2 is an oxygen-sensitive protein that controls PDH and αKGDH lipoylation and activation to support metabolic adaptation in hypoxia and cancer.
Paredes F, et al.
Proceedings of the National Academy of Sciences of the USA, 2018, 201720693-201720693 null
Poldip2 is an oxygen-sensitive protein that controls PDH and αKGDH lipoylation and activation to support metabolic adaptation in hypoxia and cancer.
Paredes F, et al.
Proceedings of the National Academy of Sciences of the USA, 115(38), 201720693-201720693 (2018)
Sirtuin lipoamidase activity is conserved in bacteria as a regulator of metabolic enzyme complexes.
Rowland E A, et al.
mBio, 8(5), e01096-e01017 (2017)
Felipe Paredes et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(8), 1789-1794 (2018-02-13)
Although the addition of the prosthetic group lipoate is essential to the activity of critical mitochondrial catabolic enzymes, its regulation is unknown. Here, we show that lipoylation of the pyruvate dehydrogenase and α-ketoglutarate dehydrogenase (αKDH) complexes is a dynamically regulated
Fotis L Kyrilis et al.
Cell reports, 34(6), 108727-108727 (2021-02-11)
The pyruvate dehydrogenase complex (PDHc) is a giant enzymatic assembly involved in pyruvate oxidation. PDHc components have been characterized in isolation, but the complex's quaternary structure has remained elusive due to sheer size, heterogeneity, and plasticity. Here, we identify fully

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service