MAK037
NAD/NADH Quantitation Kit
sufficient for 100 colorimetric tests
Synonym(s):
NAD/NADH Assay Kit
Sign Into View Organizational & Contract Pricing
All Photos(6)
About This Item
UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84
Recommended Products
usage
sufficient for 100 colorimetric tests
detection method
colorimetric
relevant disease(s)
cancer
storage temp.
−20°C
General description
Nicotinamide adenine dinucleotide (NAD) is an enzymatic cofactor involved in many redox reactions. NAD functions as an electron carrier, cycling between the oxidized (NAD) and reduced (NADH) forms. In addition to its role in redox reactions, NAD plays critical roles in ADP (adenosine diphosphate)-ribosylation reactions and as a substrate for sirtuins.
Application
NAD/NADH Quantitation Kit has been used to quantify NAD+/NADH influenced by SIRT1 (sirtuin 1) activity (associated with ischemia-reperfusion injury) in the presence of trimetazidine.
Suitability
Suitable for the detection of NADH and NAD in animal tissues (liver, kidney etc.), cell culture (adherent or suspension cells), serum or urine.
Principle
The NAD/NADH Quantification Kit provides a convenient tool for sensitive detection of the intracellular nucleotides, NAD (nicotinamide adenine dinucleotide) and NADH, and their ratio without the requirement to purify NAD/NADH from samples. This assay is specific for NAD and NADH, and does not detect NADP or NADPH. NADt (NAD and NADH) or NADH are quantified in a colorimetric assay (450 nm).
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Haiping Wang et al.
Nature immunology, 21(3), 298-308 (2020-02-19)
Depleting regulatory T cells (Treg cells) to counteract immunosuppressive features of the tumor microenvironment (TME) is an attractive strategy for cancer treatment; however, autoimmunity due to systemic impairment of their suppressive function limits its therapeutic potential. Elucidating approaches that specifically
Frédérique Moyrand et al.
Eukaryotic cell, 7(12), 2069-2077 (2008-09-30)
The genome of the basidiomycete pathogenic yeast Cryptococcus neoformans carries two UDP-glucose epimerase genes (UGE1 and UGE2). UGE2 maps within a galactose cluster composed of a galactokinase homologue gene and a galactose-1-phosphate uridylyltransferase. This clustered organization of the GAL genes
Mammalian sirtuins?emerging roles in physiology, aging, and calorie restriction.
Haigis M C and Leonard P G
Genes & Development, 20(21), 2913-2921 (2006)
Nima Borhan Fakouri et al.
Scientific reports, 7(1), 12480-12480 (2017-10-04)
Nucleic acids, which constitute the genetic material of all organisms, are continuously exposed to endogenous and exogenous damaging agents, representing a significant challenge to genome stability and genome integrity over the life of a cell or organism. Unrepaired DNA lesions
Junjun Wu et al.
Metabolic engineering, 44, 313-324 (2017-11-11)
Previous studies have made many exciting achievements on pushing the functional reversal of beta-oxidation cycle (r-BOX) to more widespread adoption for synthesis of a wide variety of fuels and chemicals. However, the redox cofactor requirement for the efficient operation of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service