M2026
Liquid scintillation vials, plastic
volume 6 mL, Minivial, cap size 18 mm , polyethylene without liner, vial size 17 mm × 57 mm , with screw cap, pkg of 1000 ea
All Photos(1)
About This Item
Recommended Products
material
high-density polyethylene vial
polypropylene cap (linerless)
feature
closure type polypropylene cap
closure type screw top vial
screw cap packed separately
packaging
pkg of 1000 ea
cap size
18 mm , polyethylene without liner
vial size
17 mm × 57 mm , with screw cap
volume
6 mL
fitting
18 thread
General description
Polyethylene with light-weight walls for increased counting efficiency.
Packaging
Vials and caps are packed in two separate plastic bags.