Skip to Content
Merck
All Photos(1)

Key Documents

H8142

Sigma-Aldrich

Hybridoma Enhancing Supplement from murine macrophage-like cell line

liquid, sterile-filtered, suitable for hybridoma

Synonym(s):

Conditioned Medium from murine macrophage-like cell line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352205

sterility

sterile-filtered

Quality Level

form

liquid

technique(s)

cell culture | hybridoma: suitable

impurities

Mycoplasma, tested
endotoxin, tested

shipped in

dry ice

storage temp.

−20°C

Application

Use at a concentration of 10% in complete medium containing FBS (10%) or CPRS-3. Optimal conditions must be determined for specific applications.

Other Notes

Conditioned medium from a proprietary murine macrophage-like cell line grown in IMDM with 2% FBS.

Caution

Avoid repeated freezing and thawing. If entire contents are not used, store frozen in working aliquots. Precipitation may occur when stored. This is not detrimental to the product and may be reversed when warmed to 37 °C.

Analysis Note

Cell line viral testing:
Murine Antibody Production (map) testing for Sendai, MHV, PVM, Reo3, GDVII, Ectomelia, MVM, Polyoma, LDMV, M. Ad, EDIM, MCMV, LCM, Hantaan.
BVD virus by fluorescent antibody stain

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service