Skip to Content
Merck
All Photos(1)

Key Documents

GE17-3712-06

HisTrap Excel

Cytiva 17-3712-06, pack of 5 × 5 mL

Synonym(s):

affinity resins, Ni Sepharose excel column, ion affinity chromatography column, IMAC column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

form

prepacked column

crosslinking

cross-linked

packaging

pack of 5 × 5 mL

manufacturer/tradename

Cytiva 17-3712-06

storage condition

(20% Ethanol)

parameter

5 bar
5 ml/min - 20 ml/min flow rate

technique(s)

liquid chromatography (LC): suitable

bed size

16 mm × 25 mm

bed volume

5 mL

column I.D.

16 mm

matrix

highly cross-linked 6% agarose

matrix active group

Nickel

average diameter

90 μm

cleaning

2-14

working range

2-13

capacity

≥10 mg binding capacity (histidine-tagged protein/ml sedimented medium)

separation technique

affinity

General description

Cleaning-in-place: pH interval where the medium can be subjected to cleaning-in-place without significant change in function.
HisTrap excel columns are prepacked with Ni Sepharose excel affinity resins for capture and purification of histidine-tagged proteins secreted into eukaryotic cell culture supernatants by immobilized metal ion affinity chromatography (IMAC). HisTrap excel 1 mL and 5 mL are ready-to-use IMAC columns prepacked with Ni Sepharose excel. The design of the columns in combination with the specific properties of the resin enables fast and convenient purifications. The special type of filter in the top and bottom of the columns makes it possible to load large volumes of cell-free, unclarified samples directly on the columns without causing back pressure problems. This time-saving property helps prevent degradation and loss of sensitive target proteins.

Application

Techniques & Applications: Ready-to-use IMAC columns, prepacked with Ni Sepharose excel medium resistant to chelating and reducing agents

Features and Benefits

  • Load eukaryotic cell culture samples containing secreted histidine-tagged proteins directly with retained binding capacity
  • Increase target protein yield and decrease degradation through reduced and simplified sample handling
  • HisTrap excel columns allow direct purification of cell-free, unclarified samples
  • HisTrap excel columns are made of biocompatible polypropylene. The columns are delivered with a stopper on the inlet and a snap-off end on the outlet.
  • HisTrap excel columns can be operated either with a peristaltic pump or with chromatography systems such as ÄKTA systems. The columns cannot be opened or refilled.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

2O at room temperature. Maximum flow rate will be lower when using buffers or samples with high viscosity or performing purification at low temperature. Optimal flow rate during binding is sample-dependent. During column wash and elution, a flow rate of 5 ml/min is recommended.
The maximum pressure the packed bed can withstand depends on the chromatography medium characteristics and sample/liquid viscosity. The value measured on the chromatography system used also depends on the tubing used to connect the column.

Legal Information

HisTrap is a trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service