Skip to Content
Merck
All Photos(1)

Documents

EHU073541

Sigma-Aldrich

MISSION® esiRNA

targeting human MGEA5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AACTGCACCTTGTGAATGGTAGTTGAGGTCTTCATACAGTTCAGCCTCTAGAATGGTAACAAATCAGCCAATTGGATTCGAAACAAAGAAGACTATGTAAAACTCACCCATCACACTTTGAGACTACTCACTGGTTGGAAGAATATAGTATTGCAGCAAATCCTGTATGAAAGAGAGATGTGGGCTTCCTTTTTGAGTCTTGTGTTAGGTGCTGAGACCTTTTACATGGGCTTATACAGGGAGAGAGTCTTCAATAAATGTAGTCAGCACTATTTTCTGCATCCAGTGTGGTTGCGTTTCTCACCTGAGAGTAATCAAGATAACATCTGTCATCTTCCTTGGTTTATTGAGTGAAATGCCTCTCAGTCTTAGGGGACATGGCAGAGATGAAAGAAAGAAAGAGTGGGTTTCAGAAGTGTCAGGGTGGAGTGATTCCAAGTGGGATGGTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel C Lucena et al.
The Journal of biological chemistry, 291(25), 12917-12929 (2016-04-30)
Deregulated cellular metabolism is a hallmark of tumors. Cancer cells increase glucose and glutamine flux to provide energy needs and macromolecular synthesis demands. Several studies have been focused on the importance of glycolysis and pentose phosphate pathway. However, a neglected
Md Ataur Rahman et al.
Oxidative medicine and cellular longevity, 2019, 6279313-6279313 (2019-12-13)
The addition of O-linked β-N-acetylglucosamine (O-GlcNAcylation) to serine and threonine residues is a common posttranslational modification of intracellular proteins which modulates protein functions and neurodegenerative diseases, controlled by a single pair of enzymes, O-GlcNAcase (OGA), and O-GlcNAcylation transferase (OGT). Autophagy
Maïté Leturcq et al.
Cellular and molecular life sciences : CMLS, 75(23), 4321-4339 (2018-08-03)
O-GlcNAcylation of proteins is governed by O-GlcNAc transferase (OGT) and O-GlcNAcase (OGA). The homeostasis of O-GlcNAc cycling is regulated during cell cycle progression and is essential for proper cellular division. We previously reported the O-GlcNAcylation of the minichromosome maintenance proteins
Anupriya Chatterjee et al.
Cells, 9(10) (2020-10-23)
Our previous studies identified that retinal endothelial damage caused by hyperglycemia or nucleoside diphosphate kinase-B (NDPK-B) deficiency is linked to elevation of angiopoietin-2 (Ang-2) and the activation of the hexosamine biosynthesis pathway (HBP). Herein, we investigated how NDPK-B is involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service