Skip to Content
Merck
All Photos(1)

Key Documents

D4072

Sigma-Aldrich

Delta Sleep Inducing Peptide

Synonym(s):

DSIP, Emideltide, Sleep inducing peptide, Trp-Ala-Gly-Gly-Asp-Ala-Ser-Gly-Glu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H48N10O15
CAS Number:
Molecular Weight:
848.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

powder

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)[C@@H](N)Cc1c[nH]c2ccccc12)C(=O)NCC(=O)NCC(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](CO)C(=O)NCC(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C35H48N10O15/c1-16(41-32(56)20(36)9-18-11-37-21-6-4-3-5-19(18)21)30(54)39-12-25(47)38-13-26(48)44-23(10-29(52)53)34(58)42-17(2)31(55)45-24(15-46)33(57)40-14-27(49)43-22(35(59)60)7-8-28(50)51/h3-6,11,16-17,20,22-24,37,46H,7-10,12-15,36H2,1-2H3,(H,38,47)(H,39,54)(H,40,57)(H,41,56)(H,42,58)(H,43,49)(H,44,48)(H,45,55)(H,50,51)(H,52,53)(H,59,60)/t16-,17-,20-,22-,23-,24-/m0/s1

InChI key

ZRZROXNBKJAOKB-GFVHOAGBSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Trp-Ala-Gly-Gly-Asp-Ala-Ser-Gly-Glu-OH

General description

Delta sleep inducing peptide (DSIP) is a nonapeptide which was isolated from the rabbit brain. It is synthesized in the hypothalamus and is present in small amounts in the blood.

Biochem/physiol Actions

DSIP promotes sleep by targeting various areas of the brain. The peptide is easily absorbed from the gut and is not denatured by enzymes. It crosses the blood-brain barrier easily. DSIP influences the circadian rhythm in humans. Low concentrations of the peptide are observed in patients with schizophrenia and depression.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delta sleep-inducing peptide
Pollard BJ and Pomfrett CJD
European Journal of Anaesthesiology, 18(7), 419-422 (2001)
Joo Sung Kim et al.
Macromolecular bioscience, 20(11), e2000147-e2000147 (2020-07-15)
Mechanoreceptors in human skin possess high sensitivity, wide sensing range, and high sensing resolution for external stimuli. Several attempts have been made to implement electronic skin (e-skin) that can mimic human skin. However, previous attempts are limited by the fundamental
Piia Liigand et al.
Journal of mass spectrometry : JMS, 54(6), 481-487 (2019-03-09)
Electrospray ionization is widely used to generate gas phase ions from a variety of molecules ranging from small ions to large proteins, while the ionization mechanism is claimed to depend on the size of the molecule. For small molecules, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service