CLS352076
Corning® Falcon® Centrifuge Tubes
capacity 175 mL, conical polypropylene, cap, plug seal screw, sterile
Synonym(s):
culture tube
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.13
Recommended Products
material
conical polypropylene
sterility
sterile
feature
cap plug seal screw
manufacturer/tradename
Corning 352076
capacity
175 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Liquid Handling
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service