Skip to Content
Merck
All Photos(1)

Documents

CLS3153

Corning® disposable spinner flasks

disposable spinner flask, polystyrene, non-treated, 500 mL, sterile, 12/cs

Synonym(s):

cell culture flasks, cell culture spinner flasks, scale up cell culture flasks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.17

material

polypropylene cap
polystyrene flask
screw top cap

sterility

sterile

feature

cap
closure type Screw Cap

packaging

case of 12

manufacturer/tradename

Corning 3153

capacity

500 mL

neck diam.

100 mm

side arm diam.

45 mm

working volume

300 mL

suitability

suitable for (scale-up cell culture; production of secreted proteins)

Looking for similar products? Visit Product Comparison Guide

General description


Convenience
The Corning disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.
Confidence
Molded from virgin polystyrene and gamma-irradiated, each spinner flask system assures a clean and sterile unit that is nonpyrogenic. No more concerns with detergent residues or contamination.
Compatibility
Made of ISO 10993 polystyrene, the vessel is comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads.
Performance
The paddle size and height is optimized for each vessel size. A unique integrated magnet provides a smooth, even rotation at required speeds on any laboratory slow speed stirrer. Heat build-up in the vessel is reduced by means of a specially designed flange that raises the vessel off the stir-plate surface.

Conclusions
  • Disposable large volume spinner flasks are equivalent to conventional glass spinner flasks for cell growth and viability
  • A variety of cell types can be cultured in large volume disposable spinner flasks
  • Disposable spinner flasks maintain equal aeration as compared to standard glass spinner flasks
  • Cultured cells from glass and disposable spinner flasks are metabolically equivalent
  • Culured cells in disposable spinner flasks product equal amounts of secreted proteins as cells cultured in traditional glass spinner flasks

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service