Recommended Products
Product Name
FRTL-5, 91030711
biological source
rat thyroid
growth mode
Adherent aggregates
karyotype
Not specified
morphology
Not specified
products
Thyroglobulin
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat thyroid
Cell Line Description
This thyroid cell line has been established from normal thyroid glands from 5 to 6-week-old Fischer rats. FRTL-5 is a clonal line of the continuously cultured FRTL cells derived by successive clonal isolation. The cells are characterised by maintaining highly differentiated thyroid features such as secretion of thyroglobulin and concentration of iodide. FRTL-5 cells have been used as assays measuring the stimulatory activity of human autoimmune immunoglobulins from patients with Graves disease. Tend to grow in three-dimensional structures, monolayer not formed.
Application
Study of thyroid cells, hormone dependency and secretion.
Culture Medium
Coon′s modified Ham′s F12 medium (or Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L Inositol) + 2mM Glutamine + 10μg/ml Insulin + 10nM Hydrocortisone + 5μg/ml Transferrin + 10ng/ml gly-his-lys acetate + 10ng/ml somatostatin + 5% Foetal Bovine Serum (FBS) + 10mU/ml TSH. As an alternative to Coon′s modified Ham′s F12, it is possible to use Kaign′s modified Ham′s F12 supplemented with 45mg/L Ascorbic acid and 18mg/L Inositol.
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service