Skip to Content
Merck
All Photos(1)

Key Documents

Myb2-32

84101003, Reactivity: Anti-myb gene products, avian

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-myb gene products, avian

technique(s)

cell culture | mammalian: suitable

isotype

IgG1, kappa

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse BALB/c spleen

Cell Line Description

The antibody recognises an antigen on the primary structure of bp37v-myb and binds at or near same site as Myb2-2 and Myb2-3. It cross reacts with v-myb gene product of 45kDa and c-myb gene product of 75kDa. The antibody recognises E26 oncogene product p135gag-myb-ets.

Immunogen

The bacterially expressed fusion protein, bp37v-myb

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service