Skip to Content
Merck
All Photos(1)

Key Documents

MDCK-SIAT1 Cell Line canine

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 5071502, canine kidney, Epithelial

Synonym(s):

MDCK-SIAT1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

canine kidney

description

Canine Cocker Spaniel Kidney Sialic Acid Over Expression

packaging

tube of 5 μg 05071502-DNA-5UG
pkg of vial of cells 05071502-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Enhanced expression of 6-linked sialic acids

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Canine Cocker Spaniel Kidney Sialic Acid Over Expression

Cell Line Description

This cell line has been derived by the stable transfection of MDCK cells with the cDNA of human 2,6-sialtransferase (SIAT1). The cells express two fold?higher amounts of 6-linked sialic acids and two fold-lower amounts of 3-linked sialic acids than parent MDCK cells. MDCK-SIAT1 cells are presented as a suitable system for testing the sensitivity of human influenza virus to neuraminidase inhibitors (NAI) due to their over expression of sialyl-α2,6-galactose moieties.

Culture Medium

DMEM + 2mM Glutamine + 1mg/ml G418 + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (85-95 %) 1:10 to 1:20 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 37°C; 5% CO2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service