Skip to Content
Merck
All Photos(1)

Key Documents

C5580

Sigma-Aldrich

CABS

≥98%

Synonym(s):

4-(Cyclohexylamino)-1-butanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H21NO3S
CAS Number:
Molecular Weight:
235.34
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98%

form

powder

pH

10.0-11.4

useful pH range

10.0-11.4

pKa (25 °C)

10.7

pKa 

10.7

solubility

water: 0.176 g/mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

O=S(CCCCNC1CCCCC1)(O)=O

InChI

1S/C10H21NO3S/c12-15(13,14)9-5-4-8-11-10-6-2-1-3-7-10/h10-11H,1-9H2,(H,12,13,14)

InChI key

XNPKNHHFCKSMRV-UHFFFAOYSA-N

Linkage

Homolog of CHES and CAPS with higher pKa and similar utility in biological systems.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T Thiel et al.
Journal of biochemical and biophysical methods, 37(3), 117-129 (1998-12-31)
Four new zwitterionic butanesulfonic acid buffers that are structurally related to four families of Good buffers were evaluated for use in biological systems. These buffers, with pKa values from 7.6 to 10.7, were compared with a variety of other buffers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service