Skip to Content
Merck
All Photos(1)

Documents

C3742

Sigma-Aldrich

Chk2 Inhibitor II hydrate

≥98% (HPLC)

Synonym(s):

2-(4-(4-Chlorophenoxy)phenyl)-1H-benzimidazole-5-carboxamide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14ClN3O2 · xH2O
CAS Number:
Molecular Weight:
363.80 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

off-white to tan

solubility

DMSO: ≥2 mg/mL (warmed)

originator

Johnson & Johnson

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C2C(N=C(C3=CC=C(OC4=CC=C(Cl)C=C4)C=C3)N2)=C1)N

InChI

1S/C20H14ClN3O2/c21-14-4-8-16(9-5-14)26-15-6-1-12(2-7-15)20-23-17-10-3-13(19(22)25)11-18(17)24-20/h1-11H,(H2,22,25)(H,23,24)

InChI key

UXGJAOIJSROTTN-UHFFFAOYSA-N

Gene Information

human ... CHEK2(11200)

Biochem/physiol Actions

Chk2 Inhibitor II is a checkpoint kinase 2 inhibitor, which controls the p53 response to DNA breaks induced by radiation, leading to apoptosis. Protection of T-cells from apoptosis implies use as an adjuvant for radiation therapy in cancer. IC50 = 15 nM; Ki = 37 nM. Chk2 Inhibitor II shows 1000-fold greater selectivity for the Chk2 serine/threonine kinase than for the Cdk1/B and CK1 kinases (for which IC50 = 12 μM and 17 μM, respectively). Chk2 Inhibitor II weakly inhibits a panel of 31 other kinases (<25% inhibition at a concentration of 10 μM and prevents apoptosis of human CD4+ and CD8+ T-cells subjected to ionizing radiation (EC50 = 3 μM and 7.6 μM, respectively).

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Oropeza et al.
Science advances, 9(26), eadf2860-eadf2860 (2023-06-30)
Cell cycle dysregulation is prerequisite for cancer formation. However, it is unknown whether the mode of dysregulation affects disease characteristics. Here, we conduct comprehensive analyses of cell cycle checkpoint dysregulation using patient data and experimental investigations. We find that ATM
Lauren E Williamson et al.
Journal of virology, 93(8) (2019-02-08)
The human B cell response to natural filovirus infections early after recovery is poorly understood. Previous serologic studies suggest that some Ebola virus survivors exhibit delayed antibody responses with low magnitude and quality. Here, we sought to study the population
Ran Guo et al.
Antioxidants (Basel, Switzerland), 11(6) (2022-06-25)
Reactive oxygen species (ROS) act as a signaling intermediate to promote cellular adaptation to maintain homeostasis by regulating autophagy during pathophysiological stress. However, the mechanism by which ROS promotes autophagy is still largely unknown. Here, we show that the ATM/CHK2/ULK1
Wenyu Zhang et al.
Cell death and differentiation, 27(2), 482-496 (2019-06-19)
Both the stress-response protein, SIRT1, and the cell cycle checkpoint kinase, CHK2, play critical roles in aging and cancer via the modulation of cellular homeostasis and the maintenance of genomic integrity. However, the underlying mechanism linking the two pathways remains
Yi Luan et al.
Science advances, 8(51), eade1846-eade1846 (2022-12-22)
Cyclophosphamide and doxorubicin lead to premature ovarian insufficiency as an off-target effect. However, their oocyte death pathway has been debated. Here, we clarified the precise mechanism of ovarian depletion induced by cyclophosphamide and doxorubicin. Dormant oocytes instead of activated oocytes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service