Skip to Content
Merck
All Photos(1)

Key Documents

B6555

Nalgene® dropper bottle with screw closure

capacity 8 mL, white bottle, white cap

Synonym(s):

bottle dropper, dropper bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

low-density polyethylene
round bottle
white bottle

Joint

Joints threaded neck (15-415)

packaging

pack of 25 ea

manufacturer/tradename

Nalgene® 2751-9025

capacity

8 mL

color

white cap

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Nalgene® dropper bottles with screw closure is an alternative to various pipetting devices.
  • The tip ensures accurate dispensing of 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time.
  • Chemically resistant to most reagents.
  • LDPE clear transparent bottles for easy sample visualization.
  • Leak-proof closure - prevents sample loss.
  • Control dispensing tip ensures a secure fit.
  • Suitable for use in biotech, diagnostic and pharmaceutical applications.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service