Skip to Content
Merck
All Photos(1)

Key Documents

A7677

Supelco

Liquid chromatography column fittings

Adapter, Luer Lock to 1/4-28 threads (E)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

material

polypropylene

grade

for chromatography

technique(s)

LPLC: suitable

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service