Skip to Content
Merck
All Photos(1)

Key Documents

A112050

Sigma-Aldrich

DMT-dA(tac) Phosphoramidite

configured for ÄKTA® and OligoPilot®

Synonym(s):

DMT-dA(tac) amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H62N7O8P
CAS Number:
Molecular Weight:
944.06
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

λ

conforms (UV/VIS Identity)

compatibility

configured for ÄKTA® and OligoPilot®

nucleoside profile

base: deoxyadenosine
base protecting group: TAC
2' protecting group: none
5' protecting group: DMT
deprotection: fast

storage temp.

-10 to -25°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4c(NC(=O)COc5ccccc5)ncnc34)(c6ccccc6)c7ccc(OC)cc7

InChI

1S/C48H54N7O8P/c1-33(2)55(34(3)4)64(61-27-13-26-49)63-41-28-44(54-32-52-45-46(50-31-51-47(45)54)53-43(56)30-59-40-16-11-8-12-17-40)62-42(41)29-60-48(35-14-9-7-10-15-35,36-18-22-38(57-5)23-19-36)37-20-24-39(58-6)25-21-37/h7-12,14-25,31-34,41-42,44H,13,27-30H2,1-6H3,(H,50,51,53,56)/t41-,42+,44+,64?/m0/s1

InChI key

INUFZOANRLMUCW-SDNJGBRXSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service