Skip to Content
Merck
All Photos(1)

Documents

14561C

SAFC

EX-CELL® VPRO Serum-Free Medium for Retinoblast Cells

Animal-protein free, without L-glutamine, liquid, suitable for cell culture, sterile-filtered

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

storage condition

protect from light

technique(s)

cell culture | mammalian: suitable

components

glucose: 6.0 g/L
HEPES: no
L-glutamine: no
NaHCO3: 1.8 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

EX-CELL VPRO is an animal-protein free, serum-free medium developed for the long-term growth of Human Embryonic Retinoblast cells (PER.C6™ and related cell lines). The cells can be subcultured directly into EX-CELL VPRO from serum-free media with little or no adaptation. PER.C6 cells can be grown as suspension cultures either in shaker flasks or roller bottles, with roller bottles being the preferred culture system. Suspension cultures can be maintained, without refeeding, for approximately 10 days and can be carried for more than 20 passages with no loss of viability.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service