Skip to Content
Merck
All Photos(1)

Key Documents

NIST869B

Column selectivity test mixture for liquid chromatography

NIST® SRM® 869b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.1 mL

manufacturer/tradename

NIST®

technique(s)

liquid chromatography (LC): suitable

application(s)

general analytical

format

multi-component solution

General description

SRM 869b is useful for characterizing liquid chromatographic (LC) column selectivity for separation of PAHs. Applications to the assessment of column selectivity for other classes of compounds such as carotene isomers have also been demonstrated. A unit of SRM 869b consists of 5 ampoules, each containing 1.1 mL of the PAH mixture. For more inforamtion, plese refer to the SDS and COA.

SRM 869B_cert

SRM 869B_SDS

Other Notes

Certified for the analytes listed below.
Benzo[a]pyrene (BaP)

Reference/Informational value is provided for the following analytes.

Dibenzo[g,p]chrysene (TBN), Phenanthro[3,4-c]phenanthrene (PhPh)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service