Skip to Content
Merck
All Photos(1)

Key Documents

NIST1647F

Priority pollutant PAHs (in acetonitrile)

NIST® SRM® 1647f

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.3 mL

manufacturer/tradename

NIST®

format

multi-component solution

General description

Each unit of NIST1647F contains five 2-mL ampoules, with each containing about 1.3 mL of an acetonitrile solution of selected PAHs.For more information, please refer to the COA and SDS.SRM 1647F_certSRM 1647F_SDS

Application

SRM Prioritypollutant PAHs isintended to serve as a calibration solution for use in chromatographic methodsfor determining polycyclic aromatic hydrocarbons (PAHs).

Features and Benefits

  • NIST certified values forselected PAHs are reported and expressed as Mass fraction (mg/kg) andConcentration (mg/L).
  • Expiration details, instructions for use and storage,are provided on the NIST certificate.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Naphthalene, Acenaphthylene, Acenaphthene, Fluorene, Phenanthrene, Anthracene, Fluoranthene, Pyrene, Benz[a]anthracene, Chrysene, Benzo[b]fluoranthene, Benzo[k]fluoranthene, Benzo[a]pyrene, Dibenz[a,h]anthracene, Benzo[ghi]perylene, Indeno[1,2,3-cd]pyrene

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service