Skip to Content
Merck
All Photos(1)

Key Documents

NIST1219

Stainless Steel

NIST® SRM® 1219, Cr 16 - Ni 2 (AISI 431)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

technique(s)

X-ray spectroscopy: suitable
optical emission spectroscopy (OES): suitable

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

SRM 1219 is in the form of a disk, which measures around 34 mm (15/16 in) in diameter and 19 mm (3/4 in) in thickness. For more information, please refer to the SDS and COA.
SRM 1219_cert
SRM 1219_SDS

Application

Standard Reference Material stainless steel intended for use in the calibration of optical emission and x-ray spectrometric methods of analysis.

Other Notes

Certified for the analytes listed below.
Carbon (C), Manganese (Mn), Phosphorus (P), Sulfur (S), Silicon (Si), Copper (Cu), Nickel (Ni), Chromium (Cr), Vanadium (V), Molybdenum (Mo), Nitrogen (N)

Reference/Informational value is provided for the following analytes.
Aluminium (Al), Boron (B), Cobalt (Co), Lead (Pb), Niobium (Nb), Tin (Sn), Titanium (Ti), Tungsten (W)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service