Recommended Products
Application
FastWoRX products are hydrophobic powders that absorb most organic compounds. With them, liquid-liquid extraction (LLE) for reaction work-ups can be replaced by a simple filtration or magnetic separation. This version of the product, FastWoRX-M, allows magnetic separation. Work-ups can be done in half the time or less. Solvent use can be reduced by 90% or more. Emulsions cannot form. Automation of FastWoRX-based work-ups is much easier than automating LLE. For more details about the product, please see our Technical Spotlight.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service