Skip to Content
Merck
All Photos(1)

Documents

90554

Millipore

UVM Listeria Selective Enrichment Broth, modified I & II

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Listeria Enrichment Medium Base, UVM, University of Vermont Medium, modified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

beef extract, 5.0 g/L
casein enzymic hydrolysate, 5.0 g/L
disodium hydrogen phosphate, 12.0 g/L
esculin, 1.0 g/L
monopotassium hydrogen phosphate, 12.0 g/L
proteose peptone, 5.0 g/L
sodium chloride, 20.0 g/L
yeast extract, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages
pathogen testing
sample preparation

microbiology

suitability

selective for Listeria spp.

Application

Listeria Enrichment Medium Base (UVM Medium) is used for selective isolation and cultivation of Listeria monocytogenes from clinical specimens.

Preparation Note

Suspend 27.17 grams in 500 ml distilled water. Heat if necessary to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Cool to 50°C and aseptically add rehydrated contents of 1 vial of Listeria UVM Supplement I (Cat. No. No. 50195) for primary enrichment or 1 vial of Listeria UVM Supplement II (Cat. No. 50279) for secondary enrichment. Mix well and dispense as desired.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service