Skip to Content
Merck
All Photos(1)

Key Documents

900521

Sigma-Aldrich

Chlorobenzene

anhydrous, ZerO2®, 99.8%

Synonym(s):

Chlorobezene ZerO2®, Degassed and low oxygen chlorobenze

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
Beilstein:
605632
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
NACRES:
NA.04
grade:
ZerO2®
bp:
132 °C (lit.)
vapor pressure:
11.8 mmHg ( 25 °C)

grade

ZerO2®

Quality Level

vapor density

3.86 (vs air)

vapor pressure

11.8 mmHg ( 25 °C)

Assay

99.8%

form

liquid

autoignition temp.

1178 °F

quality

anhydrous

expl. lim.

7.1 %

dilution

(for analytical testing)

impurities

≤0.005% water
water

refractive index

n20/D 1.524 (lit.)

pH

7

bp

132 °C (lit.)

mp

−45 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service