63014
MacConkey MUG Agar
suitable for microbiology, NutriSelect® Plus
About This Item
Recommended Products
form
powder
Quality Level
shelf life
limited shelf life, expiry date on the label
composition
agar, 13.5 g/L
bile salt mixture, 1.5 g/L
casein peptone, 17 g/L
crystal violet, 0.001 g/L
lactose, 10 g/L
meat peptone, 3 g/L
4-methylumbelliferyl-β-D-glucuronide, 0.1 g/L
neutral red, 0.03 g/L
sodium chloride, 5 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
7.1±0.2 (25 °C)
application(s)
agriculture
clinical testing
environmental
food and beverages
water monitoring
microbiology
suitability
selective and differential for Enterobacter spp.
selective and differential for Escherichia coli
selective and differential for Proteus spp.
selective and differential for Salmonella spp.
selective and differential for Shigella spp.
selective and differential for coliforms
selective and differential for enterobacteriaceae
Application
Bile salts and crystal violet extensively inhibit the Gram-positive flora. The presence of lactose and neutral red indicate lactose-positive colonies from which E. coli can be identified by fluorescence in the UV.
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service