Skip to Content
Merck
All Photos(1)

Key Documents

34104

Supelco

PCB No 11

analytical standard

Synonym(s):

3,3′-Dichlorobiphenyl, 3,3-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl2
CAS Number:
Molecular Weight:
223.10
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1cccc(c1)-c2cccc(Cl)c2

InChI

1S/C12H8Cl2/c13-11-5-1-3-9(7-11)10-4-2-6-12(14)8-10/h1-8H

InChI key

KTXUOWUHFLBZPW-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa A Rodenburg et al.
Environmental science & technology, 44(8), 2816-2821 (2010-04-14)
The non-Aroclor congener 3,3'-dichlorobiphenyl (PCB 11) has been recently detected in air, water, biota, sediment, and suspended sediment. Although it has been known since at least the 1970s that this congener is produced inadvertently during the production of diarylide yellow
Dingfei Hu et al.
Environmental science & technology, 42(21), 7873-7877 (2008-11-27)
Air samples were collected in Chicago, Illinois in 2007, and 3,3'-dichlorobiphenyl (PCB11, CAS 2050-67-1) was detected and quantified using GC/MS/MS in 91% of 184 samples. To the best of our knowledge, this is the first published report of PCB11 in
Ilora Basu et al.
Environmental science & technology, 43(17), 6488-6492 (2009-09-22)
Relatively high concentrations of 3,3'-dichlorobiphenyl (PCB-11) have been reported in water and air, and it has been suggested that this compound did not come from commercial PCB products. We report here data on atmospheric partial pressures of PCB-11 and of
T L King et al.
Marine pollution bulletin, 44(7), 590-596 (2002-09-12)
3,3'-dichlorobiphenyl (IUPAC No. 11), a chlorobiphenyl (CB) that is not generally analysed in environmental studies of CBs, is found, sometimes at high concentrations, in water, suspended particulate material, biota and sediments from Halifax Harbour, NS, Canada. The results presented demonstrate
A Kubátová et al.
Isotopes in environmental and health studies, 34(4), 325-334 (1999-03-25)
Biodegradation of polychlorinated biphenyls (PCBs) in soil is considered to be very complex due to various physico-chemical factors involved. Isotope labelling technique is the best to trace fate of the xenobiotic in the environment. In this work, the uniformly 14C-labelled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service