Skip to Content
Merck
All Photos(1)

Key Documents

31488

Supelco

Lithium tetraborate

anhydrous, puriss. p.a., ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2B4O7
CAS Number:
Molecular Weight:
169.12
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
form:
powder

grade

puriss. p.a.

Assay

≥98%

form

powder

quality

anhydrous

impurities

≤0.005% Heavy metals (as Pb)

mp

760-880 °C (lit.)

density

≥0.25 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
phosphate (PO43-): ≤20 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Na: ≤50 mg/kg

SMILES string

[Li+].[Li+].[O-]B(OB=O)OB([O-])OB=O

InChI

1S/B4O7.2Li/c5-1-9-3(7)11-4(8)10-2-6;;/q-2;2*+1

InChI key

PSHMSSXLYVAENJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hailong Hong et al.
Optics express, 21(6), 7285-7293 (2013-04-03)
We demonstrate the improvement and formation of UV-induced damage on LBO crystal output surface during long-term (130 h) high-power (20 W) high-repetition-rate (80 kHz) third-harmonic generation. The output surface was super-polished (RMS surface roughness <0.6 nm) to sub-nanometer scale super
S Miljanić et al.
Radiation protection dosimetry, 106(3), 253-256 (2003-12-24)
The aim of this work was to determine energy dependence characteristics in terms of values of the personal dose equivalent, HP(10). The following types of thermoluminescent detectors (TLDs) were investigated: (a) two new types based on lithium borate, Li2B4O7:Cu,In and
Zheng Quan et al.
Applied optics, 48(16), 2979-2982 (2009-06-03)
Efficient cw intracavity frequency doubling of a diode end-pumped Nd:GdVO4 laser that operates in the 4F(3/2) --> 4I(9/2) transition at 912 nm is demonstrated. A 15 mm long lithium borate crystal, cut for critical type I phase matching at room
P E Warwick et al.
Analytical chemistry, 73(14), 3410-3416 (2001-07-31)
Americium-241 and plutonium determinations will become of greater importance over the coming decades as 137Cs and 241Pu decay. The impact of 137Cs on environmental chronology has been great, but its potency is waning as it decays and diffuses. Having 241Am
Woo Jin Ko et al.
Journal of endourology, 26(4), 313-317 (2012-03-24)
The goal of this study is to identify the most efficient sweeping angle (SA) during photoselective vaporization of the prostate (PVP). Experiments were conducted with GreenLight XPS™ laser at 120 and 180 W. Ten blocks of porcine kidney were used for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service