Skip to Content
Merck
All Photos(2)

Documents

26293

Male Luer Lock/Removable Needle Hub Adapter

converts large hub (250 μL or larger) to a Male Luer Lock connection

Synonym(s):

Fittings for Sample Lock syringes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

description

Male Luer Lock/Removable needle hub

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 35083

compatibility

for use with Hamilton Sample Lock Syringes

Other Notes

The adapter has a port I.D. of .039" that allows for sample spiking through a syringe fitted with a 26 gauge needle or smaller.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service