Skip to Content
Merck
All Photos(1)

Key Documents

15235

Supelco

N,N-Bis(trimethylsilyl)methylamine

for GC derivatization, LiChropur, ≥97.0% (GC)

Synonym(s):

Heptamethyldisilazane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H21NSi2
CAS Number:
Molecular Weight:
175.42
Beilstein:
1737290
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥97.0% (GC)

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.421 (lit.)

bp

144-147 °C (lit.)

density

0.799 g/mL at 20 °C (lit.)

SMILES string

CN([Si](C)(C)C)[Si](C)(C)C

InChI

1S/C7H21NSi2/c1-8(9(2,3)4)10(5,6)7/h1-7H3

InChI key

ZSMNRKGGHXLZEC-UHFFFAOYSA-N

General description

N,N-Bis(trimethylsilyl)methylamine is a silylating agent used to prepare TMS derivatives. It has about the same silylation potential as hexamethyldisilazane (HMDS) and can be used in the same manner. Its reaction by-product is the gaseous methylamine.

Other Notes

Reagent for methylamination of acid halides, e.g. a phosphorus fluoride

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Meyer et al.
Chemische Berichte, 121, 627-627 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service