ZEQ7C0NDC
Feed Water Conductivity Cell
For use with Milli-Q® EQ 7000 water purification systems, monitors the ionic quality of feed water
Synonym(s):
Feed Water Conductivity Cell
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104214
NACRES:
JA.13
Recommended Products
feature
Measures the conductivity of water
manufacturer/tradename
Milli-Q®
compatibility
for use with Milli-Q® EQ 7000
Looking for similar products? Visit Product Comparison Guide
General description
The feed water conductivity cell is an installation accessory that triggers an alarm if the ionic content of feed water is out of specifications. This prevents premature exhaustion of IPAK Meta® and IPAK Quanta® polishing cartridges.
Application
The feed water conductivity cell is used to monitor the ionic quality of feed water for Milli-Q® EQ 7000 water purification systems.
Legal Information
IPAK META is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service