Skip to Content
Merck
All Photos(2)

Key Documents

PXB300C50

Millipore

Pellicon® XL Cassette with Biomax® Membrane

filtration area 50 cm2, NMWCO 300 kDa

Synonym(s):

Pellicon XL Ultrafiltration Module Biomax 300 kDa 0.005 m2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
NACRES:
NB.24

material

polyethersulfone membrane
polypropylene
polypropylene screen

Quality Level

feature

holdup volume 3.2 mL

manufacturer/tradename

Pellicon®

parameter

0.03–0.05 L/min recirculation rate
15 mL min. working volume
2.8 bar max. transmembrane pressure (40 psig)
5.5 bar max. inlet pressure (80 psi) at 50 °C

technique(s)

analytical sample preparation: suitable
protein purification: suitable

L

18.8 cm (7.4 in.)

W

3 cm (1.2 in.)

filtration area

50 cm2

membrane nominal area

50 cm2

gravimetric extractables

≤7.5 mg/device

matrix

Biomax®

pore size

300 kDa NMWCO

pH

1 –14

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Device Configuration: Cassette
Filter Code: Biomax 300
Filter Type: Ultrafiltration

Features and Benefits

Type C screen (coarse screen)

Packaging

Wet, preserved

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Analysis Note

Gravimetric Extractables: after a water flush

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service