MX0HC20CL3
Millistak+® HC Pod Depth Filter
X0HC, cellulose media, filtration area 20 cm2, hydrophilic
Synonym(s):
20 cm2 surface area, Female-Luer fitting connection, Millistak+ HC Pod Depth Filter, Millistak+ X0HC Micro 20 Depth Filter
About This Item
material
cellulose media
filter aid (inorganic)
glass filled polypropylene housing
Quality Level
feature
hydrophilic
manufacturer/tradename
Millistak+®
greener alternative product characteristics
Waste Prevention
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
diam.
2.5 in. (6.4 cm)
filtration area
20 cm2
impurities
<0.25 EU/mL (LAL test)
total organic carbon (TOC) residue
720-4600 ppb
input
sample type liquid
conductivity
3.64-10.5 μS/cm
fitting
female Luer fitting
filter grade
X0HC
greener alternative category
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Improved scalability during media screening trials
- Smaller filtration area slightly reduces feedstream volume requirements (approximately 15% less).
- Transparent housing enables visibility of fluid flow
- Easier to calculate flow rates based on 20 cm2 filter area
Millistak+® HC (high capacity) media offers:
- Millistak+® HC dual-action media improves prefiltration and compresses clarification
Millistak+® Pod filters offer:
- Low hold-up volume for greater product yield
- Broad range of media grades offered in single and multilayer products
- Flexible, modular format offers scalability from 5 to 12,000 liters or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Self-contained Pod filters protect operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Packaging
Other Notes
Legal Information
Disclaimer
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service