Skip to Content
Merck
All Photos(1)

Key Documents

MABF949

Sigma-Aldrich

Anti-Rabies Virus Antibody, clone 1112-1

clone 1112-1, from mouse

Synonym(s):

Anti-Rabies antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1112-1, monoclonal

species reactivity

rabies virus

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
neutralization: suitable

isotype

IgG1κ
IgG1

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

Glycoprotein of Evelyn Rokitniki Abelseth (ERA) SAD derived Rabies virus strain (UniProt: P03524) is encoded by the G gene. It belongs to the lyssavirus glycoprotein of the Rhabdoviridae family. This glycoprotein is a single pass type I virion membrane protein that helps in attaching the virus to host cellular receptor, inducing endocytosis of the virion. In the endosome, the acidic pH induces conformational changes in the glycoprotein trimer, which trigger fusion between virus and cell membrane. It has been shown that the muscular form of the nicotinic acetylcholine receptor (nAChR), the neuronal cell adhesion molecule (N-CAM), and the p75 neurotrophin receptor (p75NTR) bind glycoprotein and facilitate rabies virus entry into cells. Arg352 of Glycoprotein is highly involved in rabies virus pathogenicity and mutation at this site dramatically attenuates the virus.

Specificity

Clone 1112-1 detects glycoprotein of ERA strain of rabies virus in L929 cells that express rabies protein.
Clone 1112-1 detects glycoprotein of ERA strain of rabies virus in L929 cells that express rabies protein.

Immunogen

Glycoprotein of ERA (Evelyn Rokitniki Abelseth) strain Rabies virus.
Glycoprotein of ERA (Evelyn Rokitniki Abelseth) strain Rabies virus.

Application

Anti-Rabies Virus, clone 1112-1, Cat. No. MABF949, is a mouse monoclonal antibody that detects glycoprotein of strain ERA rabbies virus. It has been tested for use in Flow Cytometry and for Neutralizing.
Neutralizing Analysis: A representative lot neutralized Rabies Virus (Bunschoten, H., et. al. (1989). J Gen Virol. 70 ( Pt 2):291-8; Dietzchold, B., et. al. (1992). Proc Natl Acad Sci USA. 89(15):7252-6; Muller, T., et. al. (2009). PLoS Neql Trop Dis. 3(11):e542; Schumacher, C.L., et. al. (1989). J Clin Invest. 84(3):971-5).
Neutralizing Analysis: A representative lot neutralized Rabies Virus (Bunschoten, H., et. al. (1989). J Gen Virol. 70 ( Pt 2):291-8; Dietzchold, B., et. al. (1992). Proc Natl Acad Sci USA. 89(15):7252-6; Muller, T., et. al. (2009). PLoS Neql Trop Dis. 3(11):e542; Schumacher, C.L., et. al. (1989). J Clin Invest. 84(3):971-5).
Research Category
Inflammation & Immunology

Quality

Evaluated by Flow Cytometry in L929 expressing rabies protein.

Flow Cytometry Analysis: 1 µg of this antibody detected Rabies Virus in one million L929 cells expressing rabies protein.

Target description

58.66 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service