Skip to Content
Merck
All Photos(1)

Key Documents

8.18667

Sigma-Aldrich

2-Acrylamido-2-methylpropanesulfonic acid

(stabilised) for synthesis

Synonym(s):

2-Acrylamido-2-methylpropanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO4S
CAS Number:
Molecular Weight:
207.25
MDL number:
UNSPSC Code:
12352106
EC Index Number:
239-268-0
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.0000004 hPa ( 25 °C)

Quality Level

Assay

≥99.0% (acidimetric)

form

solid

potency

1000-2000 mg/kg LD50, oral (Rat)

mp

185 °C

solubility

soluble >500 g/L

density

1.1 g/cm3 at 15.6 °C

bulk density

640 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[S](=O)(=O)(O)CC(\N=C(/[O-])\C=C)(C)C

InChI

1S/C7H13NO4S.Na/c1-4-6(9)8-7(2,3)5-13(10,11)12;/h4H,1,5H2,2-3H3,(H,8,9)(H,10,11,12);/q;+1/p-1

InChI key

FWFUWXVFYKCSQA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service