Skip to Content
Merck
All Photos(1)

Key Documents

71147

Sigma-Aldrich

pACYCDuet-1 DNA - Novagen

pACYCDuet-1 is designed for the coexpression of two target genes. The vector encodes two multiple cloning sites (MCS) each of which is receded by a T7 promoter, lac operator and ribosome binding site.

Synonym(s):

pACYCDuet Vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106602
NACRES:
NA.51

recombinant

expressed in E. coli

Quality Level

tag

His tagged
S-Tag tagged

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze

Origin of replication

p15A

Promoter

Promoter name: T7

selection

chloramphenicol

shipped in

wet ice

storage temp.

−70°C

General description

Coexpression of multiple target genes in E. coli is advantageous for studying protein complexes. Coexpression often achieves optimal yield, solubility, and activity and may protect individual subunits from degradation. The Duet vectors are T7 promoter expression vectors, each designed to coexpress two target proteins in E. coli. The Duet vectorscarry compatible replicons and antibiotic resistance markers and may be used together in appropriate host strains to coexpress up to eight proteins. Certain combinations of Duet vectors and pET or pETcoco vectors are also compatible for coexpression. The capability of Duet vectors to be cotransformed, propagated, and induced for robust target proteincoexpression makes them ideal for the analysis of protein complexes. The Duet vectors are designed with compatible replicons and drug resistance genes for effective propagation and maintenance of four plasmids in a single cell.
pACYCDuet-1 is designed for the coexpression of two target genes. The vector encodes two multiple cloning sites (MCS) each of which is receded by a T7 promoter, lac operator and ribosome binding site (rbs). The vector also carries the P15A replicon, lacI gene and chloramphenicol resistance gene. This vector can be transformed into the same cell with pETDuet-1, pCDFDuet-1, and pRSFDuet-1 or pCOLADuet-1 in the appropriate host strain for the coexpression of up to eight target genes. Genes inserted into MCS-1 can be sequenced using the ACYCDuetUP1 Primer and DuetDOWN1 Primer. Genes inserted into MCS-2 can be sequenced using the DuetUP2 Primer and T7 Terminator Primer.




Commercial use of this Product requires a commercial license from EMD Millipore Corporation. Commercial use shall include but not be limited to (1) use of the Product or its components in manufacturing; (2) use of the Product or its components to provide a service, information, or data to others in exchange for consideration; (3) use of the Product or its components (or any derivatives thereof) for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the Product or its components, whether or not such Product or its components are resold for use in research. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of this Product.

Warning

Toxicity: Standard Handling (A)

Other Notes

Vector Family: Duet

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service