Skip to Content
Merck
All Photos(1)

Key Documents

662105

Sigma-Aldrich

Ubiquitin E1 Inhibitor, PYR-41

The Ubiquitin E1 Inhibitor, PYR-41 controls the biological activity of Ubiquitin E1. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Ubiquitin E1 Inhibitor, PYR-41, 4-(4-(5-Nitro-furan-2-ylmethylene)-3,5-dioxo-pyrazolidin-1-yl)-benzoic acid ethyl ester, trihydrate, Ubiquitin-Activating Enzyme E1 Inhibitor, PYR-41

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13N3O7 · 3H2O
Molecular Weight:
425.35
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark purple

solubility

DMSO: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable pyrazone compound that irreversibly (presumably via covalent modification) inhibits ubiquitin-activating enzyme E1 activity (IC50<10 µM in cell-free E1 ubiquitination reactions), while exhibiting little or no activity against E3, E2, or caspase enzymatic activity. Blocks ubiquitination-dependent protein degradation and other ubiquitination-mediated cellular activities. E1 inactivation by PYR-41 or other means has been shown to result in an overall elevation of sumoylation.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C. At room temperature, compound will undergo ~1% decomposition per day.

Other Notes

Yang, Y., et al. 2007. Cancer Res.67, 9472.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subhash Mehto et al.
The EMBO journal, 41(23), e111289-e111289 (2022-10-13)
The NOD1/2-RIPK2 is a key cytosolic signaling complex that activates NF-κB pro-inflammatory response against invading pathogens. However, uncontrolled NF-κB signaling can cause tissue damage leading to chronic diseases. The mechanisms by which the NODs-RIPK2-NF-κB innate immune axis is activated and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service