Skip to Content
Merck
All Photos(2)

Key Documents

559273

Sigma-Aldrich

S6K1 Inhibitor, PF-4708671

The S6K1 Inhibitor, PF-4708671 controls the biological activity of S6K1. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

S6K1 Inhibitor, PF-4708671, (2-((4-(5-Ethylpyrimidin-4-yl)piperazin-1-yl)methyl)-5-(trifluoromethyl)-1H-benzo[d]imidazole, p70 Ribosomal S6 Kinase 1 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21F3N6
Molecular Weight:
390.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL, pale yellow

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc2nc([nH]c2cc1)CN3CCN(CC3)c4ncncc4CC

General description

A cell-permeable piperazinyl-pyrimidine compound that acts as an inhibitor against S6K1 (Ki = 20 nM; IC50 = 160 nM) and MSK1 (IC50 = 950 nM) kinase activity, exhibiting little or much reduced potency against 85 other protein and lipid kinases (IC50 = 4.7, 9.2, 65 µM, respectively, against RSK1, RSK2, and S6K2; ≤27% inhibition of the rest at 1 µM). Shown to selectively inhibit IGF1- and PMA-stimulated S6K1 substrates phosphorylation in HEK-293 cells in a dose-dependent manner (up to 10 µM) in vitro and alleviate heart remodeling and functional damage in a MI (myocardial infarction) model in mice in vivo (75/mg/kg/daily i.p.).
A cell-permeable piperazinyl-pyrimidine compound that acts as an inhibitor against S6K1 (Ki = 20 nM; IC50 = 160 nM) and MSK1 (IC50 = 950 nM) kinase activity, exhibiting little or much reduced potency against a panel of 75 other protein kinases (IC50 = 4.7, 9.2, 65 µM, respectively, against RSK1, RSK2, and S6K2; ≤27% inhibition of the rest at 1 µM) and 10 lipid kinases (≤6% inhibition at 10 µM). Inhibits IGF1-induced phosphorylation of cellular S6K1 substrates S6 (Ser235/236 and Ser240/244), mTOR (Ser2448), and Rictor (Thr1135) in serum-starved HEK-293 cultures, but not the phoshorylation of MSK or RSK cellular substrates (CREB Ser133 and GSK3α/β Ser21/ Ser9, respectively) upon PMA stimulation even at concentrations as high as 10 µM. Inhibition of S6K1 activation by rapamycin (Cat. Nos. 553210, 553211, & 553212) or S6K1 activity by PF-4708671 (75/mg/kg/daily i.p.) are both shown to alleviate heart remodeling and functional damage following left coronary artery ligation-induced MI (myocardial infarction) in mice in vivo. Cellular PF-4708671 treatment is also reported to result in increased S6K1 phosphorylation, likely due to a blockage of down-stream negative feedback mechanism.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C.) Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Di, R., et al. 2012. Biochem. J.441, 199.
Pearce, L.R., et al. 2010. Biochem. J.431, 245.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna L Fricke et al.
Journal of proteome research, 22(3), 768-789 (2023-02-11)
Phosphorylation-dependent signal transduction plays an important role in regulating the functions and fate of skeletal muscle cells. Central players in the phospho-signaling network are the protein kinases AKT, S6K, and RSK as part of the PI3K-AKT-mTOR-S6K and RAF-MEK-ERK-RSK pathways. However
Taiqi Chen et al.
Nature communications, 15(1), 686-686 (2024-01-24)
Many types of tumors feature aerobic glycolysis for meeting their increased energetic and biosynthetic demands. However, it remains still unclear how this glycolytic phenomenon is achieved and coordinated with other metabolic pathways in tumor cells in response to growth stimuli.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service