Skip to Content
Merck
All Photos(1)

Key Documents

5.30796

Sigma-Aldrich

TNF-TNFR Interaction Modulator, C87

Synonym(s):

TNF-TNFR Interaction Modulator, C87, ( E)-4-(2-(4-Chloro-3-nitrophenyl)hydrazono)-3-phenyl-1-(4-phenylthiazol-2-yl)-1H-pyrazol-5(4H)-one, ( E)-4-(2-(4-Chloro-3-nitrophenyl)), TNF-TNFR Signaling Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H15ClN6O3S
CAS Number:
Molecular Weight:
502.93
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark red

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

General description

A phenylhydrazonodihydropyrazolone compound that inhibits TNFα/β-stimulated biological activity by modulating, but not abolishing, TNF-TNFR interaction via direct TNF affinity interaction (KD = 110 nM in binding study using hTNFα). Shown to prevent murine L929 cell death following hTNFα/β (1 ng/mL; Cat. Nos. 654205 & 654215) and actinomycin D (1 g/mL; Cat. Nos. 114666 & 2472-OP) treatment (EC50 = 8.73 M by MTT assay; 20 h TNF/ActD treatment) by blocking TNF/ActD-induced caspase-3/8 cleavages & IκBα degradation without affecting Fas- (CD95) mediated Jurkat cell death or VP-16 (Etoposide; Cat. No. 341205) and ADR (Adriamycin/Doxorubicin; Cat. No. 324380) cytotoxicity in L292 cultures. When applied prior to D-galactosamine (1.2 g D-GalN/kg, Cat. No. 34539) & Lipopolysaccharide (LPS; 50 g/kg) injection, C87 effectively prevents D-GalN/LPS-induced upregulation of plasma alanine transaminase & aspartate transaminase (ALT & AST) activity, resulting in significantly reduced liver damage and much improved survival rate (58.5% vs. 20.8% with or without 3X 12.5 mg C87/kg i.p. dosings 16, 8, & 1 h prior to D-GalN/LPS injection) in a murine hepatitis model in vivo. A slow and long-lasting (>12 h) JNK T183/Y185 phosphorylation is induced upon TNFα stimulation in L929 cultures, while a much faster and more transient, albeit robust, TNFα-induced JNK pT183/Y185 is seen in the presence of C87 or a TNFα neutralizing antibody.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TNFα
Reversible: yes
Target Ki: 110 nM for hTNF&alpha

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Ma, L., et al. 2014. J. Biol. Chem.289, 12457.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service