Skip to Content
Merck
All Photos(1)

Key Documents

124038

Sigma-Aldrich

Akt Inhibitor V, Triciribine

InSolution, ≥95%

Synonym(s):

InSolution Akt Inhibitor V, Triciribine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N6O4
CAS Number:
Molecular Weight:
320.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

ambient

storage temp.

−70°C

SMILES string

[n]2(c3ncnc4c3c(c2)C(=N)NN4C)[C@@H]1O[C@@H]([C@H]([C@H]1O)O)CO

InChI

1S/C13H16N6O4/c1-18-11-7-5(10(14)17-18)2-19(12(7)16-4-15-11)13-9(22)8(21)6(3-20)23-13/h2,4,6,8-9,13,20-22H,3H2,1H3,(H2,14,17)/t6-,8-,9-,13-/m1/s1

InChI key

HOGVTUZUJGHKPL-HTVVRFAVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A cell-permeable and reversible tricyclic nucleoside that selectively inhibits the cellular phosphorylation/activation of Akt1/2/3 by targeting an Akt effector molecule other than PI 3-K or PDK. Exhibits little effect towards cellular signaling pathways mediated by PKC, PKA, SGK, Stat3, p38, ERK1/2, or JNK. Shown to preferentially induce apoptosis and growth arrest in cancer cells with aberrant Akt activity both in vitro (≥60% in cell proliferation at 20 µM) and in vivo (≥80% inhibition in tumor growth in mice at 1 mg/kg/day, i.p.).

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 2 mg/312 µl solution of Akt Inhibitor V, Triciribine (Cat. No. 124012) in DMSO.

Reconstitution

Following intitial thaw, aliquot and freeze (-70°C).

Other Notes

Karst, A., et al. 2006. Cancer Res.66, 9225.
Yang, L., et al. 2004. Cancer Res.64, 4394.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Furkel et al.
Cell reports. Medicine, 2(11), 100436-100436 (2021-11-30)
Cellular morphology has the capacity to serve as a surrogate for cellular state and functionality. However, primary cardiomyocytes, the standard model in cardiovascular research, are highly heterogeneous cells and therefore impose methodological challenges to analysis. Hence, we aimed to devise

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service