Skip to Content
Merck
All Photos(1)

Documents

1.06350

Sigma-Aldrich

Sodium hydrogen sulfate monohydrate

extra pure, Technipur®

Synonym(s):

Sodium bisulfate monohydrate, Sodium hydrate hydrogen sulfate, Sodium hydrogen sulfate-1-hydrate, Sodium hydrogensulfate hydrate, Sodium hydrogen sulfate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHSO4 · H2O
CAS Number:
Molecular Weight:
138.08
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

Quality Level

pH

1 (20 °C, 50 g/L in H2O)

mp

183 °C (decomposition)

SMILES string

[Na+].[H]O[H].OS([O-])(=O)=O

InChI

1S/Na.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+1;;/p-1

InChI key

JXHZRQHZVYDRGX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium hydrogen sulfate monohydrate can be used as a component of:
  • An electrolytic system used to prepare nanotube arrays via anodization of titanium foils for hydrogen gas sensing applications.
  • The mixture of phosphates of copper and sodium, which can be used as a heat storage material.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and thermal property of {Cu3(PO4)2?2H2O; Na3PO4; NaHSO4?H2O}
Swain T
Solar Energy, 159, 369-374 (2018)
Unprecedented ultra-high hydrogen gas sensitivity in undoped titania nanotubes
Paulose M, et al.
Nanotechnology, 17(2), 398-398 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service