100107
4-Bromo-calcium Ionophore A23187
≥90% (HPLC), solid, cation ionophore, Calbiochem®
Synonym(s):
4-Bromo-A23187, 4-Bromo-calcimycin
About This Item
Recommended Products
Product Name
4-Bromo-calcium Ionophore A23187, Brominated analog of the widely used cation ionophore A23187. Non-fluorescent and therefore suitable for use in the presence of fluorescent or metal-chelating probes.
Quality Level
Assay
≥90% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
protect from light
color
yellow
solubility
DMSO: 100 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
CNc1c(Br)cc2oc(C[C@H]3O[C@@]4(CC[C@H]3C)O[C@@H]([C@H](C)C[C@H]4C)[C@H](C)C(=O)c5ccc[nH]5)nc2c1C(O)=O
InChI
1S/C29H36BrN3O6/c1-14-8-9-29(16(3)11-15(2)27(39-29)17(4)26(34)19-7-6-10-32-19)38-20(14)13-22-33-25-21(37-22)12-18(30)24(31-5)23(25)28(35)36/h6-7,10,12,14-17,20,27,31-32H,8-9,11,13H2,1-5H3,(H,35,36)/t14-,15-,16-,17-,20-,27+,29+/m1/s1
InChI key
LDXQFZZGVTWFCF-FHYGWRBKSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Cation ionophore
Warning
Reconstitution
Other Notes
Deber, C.M., et al. 1985. Anal. Biochem.146, 349.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service