Skip to Content
Merck
All Photos(1)

Key Documents

880226P

Avanti

DSPE-PEG(5000) DBCO

Avanti Research - A Croda Brand 880226P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-n-[dibenzocycooctyl(polyethylene glycol)-5000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 5 mg (880226P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880226P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC([(OCH2CH2)113]NC(CCC(N1CC(C=CC=C2)=C2C#CC3=C1C=CC=C3)=O)=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+]

General description

DSPE-PEG(5000) DBCO, also known as 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-n-[dibenzocyclooctyl(polyethylene glycol)-5000], belongs to a group of functionalized polyethylene glycol (PEG) lipids.

Packaging

5 mL Amber Glass Screw Cap Vial (880226P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service